Tisk…

5-Hydroxynicotinaldehyde

Dodavatel: Apollo Scientific
Varování

OR53003-1G OR53003-500MG
APOSOR53003-1GEA 7710 CZK
APOSOR53003-1G APOSOR53003-500MG
5-Hydroxynicotinaldehyde
5-Hydroxynicotinaldehyde
Vzorec: C₆H₅NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13189119
CAS číslo: 1060804-48-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR