Methyl 4-chloro-3-sulphamoylbenzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR52995-1G
OR52995-250MG
OR52995-5G
APOSOR52995-1GEA
3870
CZK
APOSOR52995-1G
APOSOR52995-250MG
APOSOR52995-5G
Methyl 4-chloro-3-sulphamoylbenzoate
Methyl 4-chloro-3-sulphamoylbenzoate
Vzorec:
C₈H₈ClNO₄S MW: 249,671 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02676058 CAS číslo: 1208-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...