Tisk…

Methyl 4-bromo-3-sulphamoylbenzoate

Dodavatel: Apollo Scientific
Varování

OR52994-1G OR52994-250MG OR52994-5G
APOSOR52994-1GEA 14060 CZK
APOSOR52994-1G APOSOR52994-250MG APOSOR52994-5G
Methyl 4-bromo-3-sulphamoylbenzoate
Methyl 4-bromo-3-sulphamoylbenzoate
Vzorec: C₈H₈BrNO₄S
MW: 294,122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18205814
CAS číslo: 74451-73-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR