Tisk…

Methyl 3-chloro-4-sulphamoylbenzoate

Dodavatel: Apollo Scientific
Varování

OR52992-1G OR52992-250MG
APOSOR52992-1GEA 16340 CZK
APOSOR52992-1G APOSOR52992-250MG
Methyl 3-chloro-4-sulphamoylbenzoate
Methyl 3-chloro-4-sulphamoylbenzoate
Vzorec: C₈H₈ClNO₄S
MW: 249,671 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602571
CAS číslo: 1375473-97-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR