4-Chloro-3-(dimethylsulphamoyl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR52988-1G
OR52988-250MG
APOSOR52988-1GEA
4200
CZK
APOSOR52988-1G
APOSOR52988-250MG
4-Chloro-3-(dimethylsulphamoyl)benzoic acid
4-Chloro-3-(dimethylsulphamoyl)benzoic acid
Vzorec:
C₉H₁₀ClNO₄S MW: 263,698 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02675789 CAS číslo: 59210-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...