5-Iodo-3,4-dihydro-2H-1,8-naphthyridine, N1-BOC protected 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl 5-iodo-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylate
OR52891-1G
OR52891-250MG
OR52891-5G
APOSOR52891-1GEA
4770
CZK
APOSOR52891-1G
APOSOR52891-250MG
APOSOR52891-5G
5-Iodo-3,4-dihydro-2H-1,8-naphthyridine, N1-BOC protected 95%
5-Iodo-3,4-dihydro-2H-1,8-naphthyridine, N1-BOC protected
Vzorec:
C₁₃H₁₇IN₂O₂ MW: 360,191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31745691 CAS číslo: 2367002-56-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...