4-(1-Piperidyl)pyridin-3-amine 95%
Dodavatel: Apollo Scientific
OR52772-1G
OR52772-250MG
APOSOR52772-1GEA
21730
CZK
APOSOR52772-1G
APOSOR52772-250MG
4-(1-Piperidyl)pyridin-3-amine 95%
4-(1-Piperidyl)pyridin-3-amine
Vzorec:
C₁₀H₁₅N₃ Storage Temperature: Ambient |
MDL Number:
MFCD10691294 CAS číslo: 52311-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...