3-Piperazin-1-ylaniline 98%
Dodavatel: Apollo Scientific
OR52765-1G
OR52765-250MG
APOSOR52765-1GEA
19440
CZK
APOSOR52765-1G
APOSOR52765-250MG
3-Piperazin-1-ylaniline 98%
3-Piperazin-1-ylaniline
Vzorec:
C₁₀H₁₅N₃ MW: 177,246 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03444414 CAS číslo: 125422-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...