4-(2-Bromophenyl)morpholine 97%
Dodavatel: Apollo Scientific
OR52755-1G
OR52755-5G
APOSOR52755-1GEA
1120
CZK
APOSOR52755-1G
APOSOR52755-5G
4-(2-Bromophenyl)morpholine 97%
4-(2-Bromophenyl)morpholine
Vzorec:
C₁₀H₁₂BrNO MW: 242.112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03086182 CAS číslo: 87698-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...