Tisk…

6-Bromo-3-methybenzofuran

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Bromo-3-methylbenzofuran

OR52747-1G OR52747-250MG
APOSOR52747-1GEA 5920 CZK
APOSOR52747-1G APOSOR52747-250MG
6-Bromo-3-methybenzofuran
6-Bromo-3-methybenzofuran
Vzorec: C₉H₇BrO
MW: 211,055 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24387451
CAS číslo: 33118-86-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR