Tisk…

3-Bromo-2-iodobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-bromo-3-(bromomethyl)-2-iodobenzene

OR52736-1G OR52736-250MG OR52736-5G
APOSOR52736-1GEA 14880 CZK
APOSOR52736-1G APOSOR52736-250MG APOSOR52736-5G
3-Bromo-2-iodobenzyl bromide 95%
3-Bromo-2-iodobenzyl bromide
Vzorec: C₇H₅Br₂I
MW: 375,827 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391669
CAS číslo: 1261757-20-3
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR