Tisk…

3-Bromo-2-iodobenzyl alcohol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-bromo-2-iodophenyl)methanol

OR52730-1G OR52730-250MG OR52730-5G
APOSOR52730-1GEA 5720 CZK
APOSOR52730-1G APOSOR52730-250MG APOSOR52730-5G
3-Bromo-2-iodobenzyl alcohol 97%
3-Bromo-2-iodobenzyl alcohol
Vzorec: C₇H₆BrIO
MW: 312,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391655
CAS číslo: 1261571-03-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR