Methyl 3-bromo-2-iodobenzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR52724-1G
OR52724-250MG
OR52724-5G
APOSOR52724-1GEA
6120
CZK
APOSOR52724-1G
APOSOR52724-250MG
APOSOR52724-5G
Methyl 3-bromo-2-iodobenzoate 95%
Methyl 3-bromo-2-iodobenzoate
Vzorec:
C₈H₆BrIO₂ MW: 340,941 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398801 CAS číslo: 121772-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...