Tisk…

3-Bromo-2-nitrobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Bromo-3-(bromomethyl)-2-nitrobenzene

OR52712-1G OR52712-250MG OR52712-5G
APOSOR52712-1GEA 992 CZK
APOSOR52712-1G APOSOR52712-250MG APOSOR52712-5G
3-Bromo-2-nitrobenzyl bromide 95%
3-Bromo-2-nitrobenzyl bromide
Vzorec: C₇H₅Br₂NO₂
MW: 294.928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390264
CAS číslo: 63471-25-0
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR