Tisk…

Pyridine-4-sulphonic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Pyridinesulphonic acid, 4-Sulphopyridine

OR5271-1G OR5271-250MG
APOSOR5271-1GEA 4770 CZK
APOSOR5271-1G APOSOR5271-250MG
Pyridine-4-sulphonic acid
Pyridine-4-sulphonic acid
Vzorec: C₅H₅NO₃S
MW: 159,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234082
CAS číslo: 5402-20-0
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR