Tisk…

2,5-Dihydroxypyridine

Dodavatel: Apollo Scientific

OR52588-1G OR52588-5G
APOSOR52588-1GEA 3310 CZK
APOSOR52588-1G APOSOR52588-5G
2,5-Dihydroxypyridine
2,5-Dihydroxypyridine
Vzorec: C₅H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00234075
CAS číslo: 5154-01-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR