Tisk…

Tetrahydropyran-3-boronic acid, pinacol ester 98%

Dodavatel: Apollo Scientific
Varování

OR52587-1G OR52587-250MG OR52587-5G
APOSOR52587-1GEA 6850 CZK
APOSOR52587-1G APOSOR52587-250MG APOSOR52587-5G
Tetrahydropyran-3-boronic acid, pinacol ester 98%
Tetrahydropyran-3-boronic acid, pinacol ester
Vzorec: C₁₁H₂₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD09997791
CAS číslo: 1391850-39-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR