5-(Triethoxysilyl)pentanoic acid, tech
Dodavatel: Apollo Scientific
|
Varování
|
OR52572-100MG
OR52572-1G
OR52572-250MG
APOSOR52572-100MGEA
3580
CZK
APOSOR52572-100MG
APOSOR52572-1G
APOSOR52572-250MG
5-(Triethoxysilyl)pentanoic acid, tech
5-(Triethoxysilyl)pentanoic acid
Vzorec:
C₁₁H₂₄O₅Si Storage Temperature: Ambient |
MDL Number:
MFCD29067261 CAS číslo: 1137665-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...