Tisk…

tert-Butyl 4-hydroxybenzoate

Dodavatel: Apollo Scientific
Varování

OR52549-1G OR52549-25G OR52549-5G
APOSOR52549-1GEA 695 CZK
APOSOR52549-1G APOSOR52549-25G APOSOR52549-5G
tert-Butyl 4-hydroxybenzoate
tert-Butyl 4-hydroxybenzoate
Vzorec: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD09953072
CAS číslo: 25804-49-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR