Tisk…

3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile 95+%

Dodavatel: Apollo Scientific

Synonyms: 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carbonitrile, 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carbonitrile

OR52436-250MG OR52436-1G
APOSOR52436-250MGEA 2200 CZK
APOSOR52436-250MG APOSOR52436-1G
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile 95+%
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile
Vzorec: C₉H₆N₂O₂
MW: 174,156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20039908
CAS číslo: 134997-74-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR