Tisk…

Ethyl 5-bromo-1H-imidazole-4-carboxylate

Dodavatel: Apollo Scientific
Varování

OR52419-1G OR52419-250MG
APOSOR52419-1GEA 40670 CZK
APOSOR52419-1G APOSOR52419-250MG
Ethyl 5-bromo-1H-imidazole-4-carboxylate
Ethyl 5-bromo-1H-imidazole-4-carboxylate
Vzorec: C₆H₇BrN₂O₂
MW: 219,036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697506
CAS číslo: 944906-76-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR