2-Chloro-6-iodobenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR52403-1G
OR52403-5G
APOSOR52403-1GEA
7480
CZK
APOSOR52403-1G
APOSOR52403-5G
2-Chloro-6-iodobenzaldehyde
2-Chloro-6-iodobenzaldehyde
Vzorec:
C₇H₄ClIO Storage Temperature: Ambient |
MDL Number:
MFCD18393320 CAS číslo: 51738-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...