Tisk…

6-Bromo-2(1H)-quinazolinone

Dodavatel: Apollo Scientific
Varování

OR52372-1G OR52372-5G
APOSOR52372-1GEA 9460 CZK
APOSOR52372-1G APOSOR52372-5G
6-Bromo-2(1H)-quinazolinone
6-Bromo-2(1H)-quinazolinone
Vzorec: C₈H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09999188
CAS číslo: 79885-37-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR