Tisk…

(3R)-Morpholine-3-carboxylic acid hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: (R)-Morpholine-3-carboxylic acid hydrochloride

OR52357-1G OR52357-250MG
APOSOR52357-1GEA 1290 CZK
APOSOR52357-1G APOSOR52357-250MG
(3R)-Morpholine-3-carboxylic acid hydrochloride
(3R)-Morpholine-3-carboxylic acid hydrochloride
Vzorec: C₅H₉NO₃·ClH
MW: 167,591 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06809588
CAS číslo: 1187928-88-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR