N-(3-Trimethoxysilylpropyl)pyrrole 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR52338-5G
OR52338-1G
APOSOR52338-5GEA
5430
CZK
APOSOR52338-5G
APOSOR52338-1G
N-(3-Trimethoxysilylpropyl)pyrrole 95%
N-(3-Trimethoxysilylpropyl)pyrrole
Vzorec:
C₁₀H₁₉NO₃Si Bod varu: 105…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00054844 CAS číslo: 80906-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...