1,2,4-Oxadiazole-3-carboxylic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR52319-100MG
OR52319-250MG
OR52319-1G
APOSOR52319-100MGEA
3740
CZK
APOSOR52319-100MG
APOSOR52319-250MG
APOSOR52319-1G
1,2,4-Oxadiazole-3-carboxylic acid 95+%
1,2,4-Oxadiazole-3-carboxylic acid
Vzorec:
C₃H₂N₂O₃ MW: 114,06 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09414720 CAS číslo: 856787-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...