Tisk…

Methyl 3-hydroxyazetidine-1-carboxylate

Dodavatel: Apollo Scientific
Varování

OR52278-10G OR52278-1G OR52278-5G
APOSOR52278-10GEA 24270 CZK
APOSOR52278-10G APOSOR52278-1G APOSOR52278-5G
Methyl 3-hydroxyazetidine-1-carboxylate
Methyl 3-hydroxyazetidine-1-carboxylate
Vzorec: C₅H₉NO₃
MW: 131,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072727
CAS číslo: 118972-97-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR