7-Mercapto-4-methylcoumarin 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Methyl-7-thioumbelliferone, 4-Methyl-7-sulphanyl-2H-chromen-2-one, 7-Mercapto-4-methyl-2H-chromen-2-one
OR52240-1G
OR52240-5G
APOSOR52240-1GEA
1450
CZK
APOSOR52240-1G
APOSOR52240-5G
7-Mercapto-4-methylcoumarin 97%
7-Mercapto-4-methylcoumarin
Vzorec:
C₁₀H₈O₂S MW: 192,234 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00467591 CAS číslo: 137215-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...