Tisk…

3-Bromo-5-chloro-N-methoxy-N-methylbenzamide

Dodavatel: Apollo Scientific
Varování

OR52231-1G OR52231-5G
APOSOR52231-1GEA 12570 CZK
APOSOR52231-1G APOSOR52231-5G
3-Bromo-5-chloro-N-methoxy-N-methylbenzamide
3-Bromo-5-chloro-N-methoxy-N-methylbenzamide
Vzorec: C₉H₉BrClNO₂
MW: 278,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28384163
CAS číslo: 1700202-83-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR