Methyl 5-amino-3-chloropyridine-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 5-amino-3-chloropicolinate, 5-Amino-3-chloro-2-(methoxycarbonyl)pyridine
OR52216-100MG
OR52216-250MG
OR52216-1G
APOSOR52216-100MGEA
2400
CZK
APOSOR52216-100MG
APOSOR52216-250MG
APOSOR52216-1G
Methyl 5-amino-3-chloropyridine-2-carboxylate
Methyl 5-amino-3-chloropyridine-2-carboxylate
Vzorec:
C₇H₇ClN₂O₂ MW: 186,596 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18258038 CAS číslo: 1256825-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...