Tisk…

[(E)-2-(Ethoxycarbonyl)vinyl]boronic acid, pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

OR52213-1G OR52213-5G OR52213-25G
APOSOR52213-1GEA 0 CZK
APOSOR52213-1G APOSOR52213-5G APOSOR52213-25G
[(E)-2-(Ethoxycarbonyl)vinyl]boronic acid, pinacol ester 97%
[(E)-2-(Ethoxycarbonyl)vinyl]boronic acid, pinacol ester
Vzorec: C₁₁H₁₉BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD09027313
CAS číslo: 1009307-13-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR