Tisk…

5-Amino-1,3-benzoxazole hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,3-Benzoxazol-5-amine hydrochloride

OR52195-1G OR52195-5G
APOSOR52195-1GEA 3740 CZK
APOSOR52195-1G APOSOR52195-5G
5-Amino-1,3-benzoxazole hydrochloride
5-Amino-1,3-benzoxazole hydrochloride
Vzorec: C₇H₆N₂O·ClH
MW: 170,596 g/mol
Bod tání: 226…229 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28954649
CAS číslo: 1858249-87-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR