Tisk…

2-[(Tetrahydro-2H-pyran-4-yl)oxy]aniline 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(2-Aminophenoxy)tetrahydro-2H-pyran

OR5214-1G OR5214-250MG
APOSOR5214-1GEA 10780 CZK
APOSOR5214-1G APOSOR5214-250MG
2-[(Tetrahydro-2H-pyran-4-yl)oxy]aniline 95%
2-[(Tetrahydro-2H-pyran-4-yl)oxy]aniline
Vzorec: C₁₁H₁₅NO₂
MW: 193,242 g/mol
Bod tání: 58…60,5 °C
Storage Temperature: Ambient
MDL Number: MFCD09025872
CAS číslo: 898289-35-5
UN: 2811
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR