Tisk…

3-Iodo-2-nitroaniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-6-iodonitrobenzene

OR52139-1G OR52139-250MG OR52139-5G
APOSOR52139-1GEA 5460 CZK
APOSOR52139-1G APOSOR52139-250MG APOSOR52139-5G
3-Iodo-2-nitroaniline
3-Iodo-2-nitroaniline
Vzorec: C₆H₅IN₂O₂
MW: 264.021 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18397930
CAS číslo: 721925-17-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR