6-Bromo-2-chloro-7-methyl-3H-imidazo[4,5-b]pyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-2-chloro-4-methyl-7-azabenzimidazole
OR52096-1G
OR52096-250MG
APOSOR52096-1GEA
28440
CZK
APOSOR52096-1G
APOSOR52096-250MG
6-Bromo-2-chloro-7-methyl-3H-imidazo[4,5-b]pyridine
6-Bromo-2-chloro-7-methyl-3H-imidazo[4,5-b]pyridine
Vzorec:
C₇H₅BrClN₃ MW: 246,492 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22460484 CAS číslo: 1388071-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...