Tisk…

[6-(Thien-2-yl)pyridin-3-yl]methylamine dihydrochloride 90+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-[5-(Aminomethyl)pyridin-2-yl]thiophene dihydrochloride, 5-(Aminomethyl)-2-(thien-2-yl)pyridine dihydrochloride

OR5209-1G OR5209-250MG
APOSOR5209-1GEA 10720 CZK
APOSOR5209-1G APOSOR5209-250MG
[6-(Thien-2-yl)pyridin-3-yl]methylamine dihydrochloride 90+%
[6-(Thien-2-yl)pyridin-3-yl]methylamine dihydrochloride
Vzorec: C₁₀H₁₀N₂S·2ClH
MW: 263,187 g/mol
Bod tání: 270…272 °C
Storage Temperature: Ambient
MDL Number: MFCD09025878
CAS číslo: 913830-32-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR