2-(4-Methylhomopiperazin-1-yl)benzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5208-1G
OR5208-250MG
OR5208-5G
APOSOR5208-1GEA
10490
CZK
APOSOR5208-1G
APOSOR5208-250MG
APOSOR5208-5G
2-(4-Methylhomopiperazin-1-yl)benzylamine 97%
2-(4-Methylhomopiperazin-1-yl)benzylamine
Vzorec:
C₁₃H₂₁N₃ Bod varu: 122 °C (0,1 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD09065001 CAS číslo: 915707-56-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...