3-Iodo-2-nitrophenol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Hydroxy-6-iodonitrobenzene
OR52072-1G
OR52072-250MG
OR52072-5G
APOSOR52072-1GEA
3970
CZK
APOSOR52072-1G
APOSOR52072-250MG
APOSOR52072-5G
3-Iodo-2-nitrophenol
3-Iodo-2-nitrophenol
Vzorec:
C₆H₄INO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD18398038 CAS číslo: 861010-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...