Tisk…

6-Ethyl-5-iodopyrimidine-2,4-diamine 97%

Dodavatel: Apollo Scientific

Synonyms: 2,4-Diamino-6-ethyl-5-iodopyrimidine, 2,4-Diamino-6-ethyl-5-iodo-1,3-diazine

OR52066-250MG OR52066-1G OR52066-5G
APOSOR52066-250MGEA 4400 CZK
APOSOR52066-250MG APOSOR52066-1G APOSOR52066-5G
6-Ethyl-5-iodopyrimidine-2,4-diamine 97%
6-Ethyl-5-iodopyrimidine-2,4-diamine
Vzorec: C₆H₉IN₄
MW: 264,067 g/mol
Bod tání: 150…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11655943
CAS číslo: 514854-13-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR