Tisk…

3-Bromo-5-isopropylbenzoic acid

Dodavatel: Apollo Scientific
Varování

OR51942-250MG OR51942-1G
APOSOR51942-250MGEA 1740 CZK
APOSOR51942-250MG APOSOR51942-1G
3-Bromo-5-isopropylbenzoic acid
3-Bromo-5-isopropylbenzoic acid
Vzorec: C₁₀H₁₁BrO₂
MW: 243,097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20040502
CAS číslo: 112930-39-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR