4,5-Dihydro-2-hydrazino-1H-imidazole hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR51941-1G
OR51941-100MG
OR51941-250MG
OR51941-500MG
OR51941-2.5G
OR51941-5G
APOSOR51941-1GEA
3380
CZK
APOSOR51941-1G
APOSOR51941-100MG
APOSOR51941-250MG
APOSOR51941-500MG
APOSOR51941-2.5G
APOSOR51941-5G
4,5-Dihydro-2-hydrazino-1H-imidazole hydrochloride
4,5-Dihydro-2-hydrazino-1H-imidazole hydrochloride
Vzorec:
C₃H₈N₄·ClH MW: 136,583 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28987620 CAS číslo: 49541-78-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...