3-Bromo-6-chloro-2-methoxypyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR51871-1G
OR51871-5G
OR51871-10G
APOSOR51871-1GEA
3190
CZK
APOSOR51871-1G
APOSOR51871-5G
APOSOR51871-10G
3-Bromo-6-chloro-2-methoxypyridine 98%
3-Bromo-6-chloro-2-methoxypyridine
Vzorec:
C₆H₅BrClNO MW: 222,467 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18257579 CAS číslo: 1211526-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...