2-Amino-3-methylbenzyl alcohol 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Amino-3-methylphenyl)methanol
OR51830-25G
OR51830-5G
APOSOR51830-25GEA
15340
CZK
APOSOR51830-25G
APOSOR51830-5G
2-Amino-3-methylbenzyl alcohol 97%
2-Amino-3-methylbenzyl alcohol
Vzorec:
C₈H₁₁NO MW: 137.179 g/mol Bod tání: 67…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075027 CAS číslo: 57772-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...