Tisk…

2-Bromo-5-ethynylthiophene 95%

Dodavatel: Apollo Scientific
Varování

OR51798-1G OR51798-5G
APOSOR51798-1GEA 4040 CZK
APOSOR51798-1G APOSOR51798-5G
2-Bromo-5-ethynylthiophene 95%
2-Bromo-5-ethynylthiophene
Vzorec: C₆H₃BrS
MW: 187,057 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08703509
CAS číslo: 105995-73-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR