Tisk…

5-Bromo-3-hydroxy-2-methoxypyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-methoxypyridin-3-ol, 5-Bromo-3-hydroxypyridin-2-yl methyl ether

OR51784-100MG OR51784-500MG
APOSOR51784-100MGEA 2480 CZK
APOSOR51784-100MG APOSOR51784-500MG
5-Bromo-3-hydroxy-2-methoxypyridine
5-Bromo-3-hydroxy-2-methoxypyridine
Vzorec: C₆H₆BrNO₂
MW: 204,021 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256269
CAS číslo: 1211589-04-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR