Ethyl 5-hydroxynicotinate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(Ethoxycarbonyl)-5-hydroxypyridine, Ethyl 5-hydroxypyridine-3-carboxylate
OR51770-1G
OR51770-250MG
APOSOR51770-1GEA
1240
CZK
APOSOR51770-1G
APOSOR51770-250MG
Ethyl 5-hydroxynicotinate
Ethyl 5-hydroxynicotinate
Vzorec:
C₈H₉NO₃ MW: 167,162 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00510027 CAS číslo: 59288-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...