Tisk…

4-Hydroxypyridine-2,6-dicarboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Helidaminic acid, 2,6-Dicarboxy-4-hydroxypyridine, 4-Hydroxydipicolinic acid

OR51769-25G OR51769-5G OR51769-1G
APOSOR51769-25GEA 3870 CZK
APOSOR51769-25G APOSOR51769-5G APOSOR51769-1G
4-Hydroxypyridine-2,6-dicarboxylic acid 97%
4-Hydroxypyridine-2,6-dicarboxylic acid
Vzorec: C₇H₅NO₅
MW: 183.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066478
CAS číslo: 499-51-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR