Tisk…

Triphenyl-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]silane 98%

Dodavatel: Apollo Scientific

Synonyms: Triphenyl(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)silane

OR51032-1G OR51032-5G
APOSOR51032-1GEA 787 CZK
APOSOR51032-1G APOSOR51032-5G
Triphenyl-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]silane 98%
Triphenyl-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]silane
Vzorec: C₃₀H₃₁BO₂Si
MW: 462,462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32708892
CAS číslo: 1391041-75-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR