5,9-Dibromo-7,7-diphenyl-fluoreno[4,3-b]benzofuran 99%
Dodavatel: Apollo Scientific
OR51031-1G
OR51031-5G
APOSOR51031-1GEA
2690
CZK
APOSOR51031-1G
APOSOR51031-5G
5,9-Dibromo-7,7-diphenyl-fluoreno[4,3-b]benzofuran 99%
5,9-Dibromo-7,7-diphenyl-fluoreno[4,3-b]benzofuran
Vzorec:
C₃₁H₁₈Br₂O Storage Temperature: Ambient |
CAS číslo:
1872364-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...