Tisk…

5-Bromopyridazin-4-ol ≥98%

Dodavatel: Apollo Scientific

OR510142-100MG OR510142-250MG OR510142-1G
APOSOR510142-100MGEA 6550 CZK
APOSOR510142-100MG APOSOR510142-250MG APOSOR510142-1G
5-Bromopyridazin-4-ol ≥98%
5-Bromopyridazin-4-ol
Vzorec: C₄H₃BrN₂O
MW: 174.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28650545
CAS číslo: 1784428-43-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR